Boc-(S)-3-amino-4-(3-cyanophenyl)butyric acid 98%

Synonyme: (S)-3-((tert-Butoxycarbonyl)amino)-4-(3-cyanophenyl)butanoic acid

BD16452-100MG BD16452-1G BD16452-250MG
BLDPBD16452-100MGEA 113 EUR
BLDPBD16452-100MG BLDPBD16452-1G BLDPBD16452-250MG
Boc-(S)-3-amino-4-(3-cyanophenyl)butyric acid 98%
Boc-(S)-3-amino-4-(3-cyanophenyl)butyric acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₆H₂₀N₂O₄
Molecular Weight: 304,35 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD01861076
CAS-Nummer: 270065-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR