5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%

Synonyme: 5-Chloro-2-methoxypyridine-3-boronic acid pinacol ester

BD164510-5G BD164510-250MG BD164510-1G BD164510-100MG
BLDPBD164510-5GEA 617 EUR
BLDPBD164510-5G BLDPBD164510-250MG BLDPBD164510-1G BLDPBD164510-100MG
5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%
5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₁₇BClNO₃
Lagertemperatur: Refrigerator
MDL: MFCD09878898
CAS-Nummer: 1083168-96-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR