2-Methoxy-3-nitro-5-pyridinboronsäurepinakolester 95%

Synonyme: 2-Methoxy-3-nitropyridine-5-boronic acid pinacol ester,2-Methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

BD164509-100MG
BLDPBD164509-100MGEA 27 EUR
BLDPBD164509-100MG
2-Methoxy-3-nitro-5-pyridinboronsäurepinakolester 95%
2-Methoxy-3-nitro-5-pyridinboronsäurepinakolester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₁₇BN₂O₅
Molecular Weight: 280,08 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD12923397
CAS-Nummer: 1083168-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR