(2-Oxo-2,3-dihydrobenzo[d]oxazol-6-yl)boronic acid 96%

BD164465-1G BD164465-50MG BD164465-100MG BD164465-250MG
BLDPBD164465-1GEA 886 EUR
BLDPBD164465-1G BLDPBD164465-50MG BLDPBD164465-100MG BLDPBD164465-250MG
(2-Oxo-2,3-dihydrobenzo[d]oxazol-6-yl)boronic acid 96%
(2-Oxo-2,3-dihydrobenzo[d]oxazol-6-yl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₆BNO₄
Molecular Weight: 178,94 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD18072745
CAS-Nummer: 1016644-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR