2,6-Difluor-3-hydroxyphenylboronsäure 97%

Synonyme: (2,6-Difluoro-3-hydroxyphenyl)boronic acid

BD164464-250MG BD164464-1G BD164464-5G
BLDPBD164464-250MGEA 31 EUR
BLDPBD164464-250MG BLDPBD164464-1G BLDPBD164464-5G
2,6-Difluor-3-hydroxyphenylboronsäure 97%
2,6-Difluor-3-hydroxyphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₅BF₂O₃
Molecular Weight: 173,91 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09878345
CAS-Nummer: 957065-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR