2-Amino-5-methoxypyrimidine 97%

BD164456-250MG BD164456-1G BD164456-5G BD164456-100MG
BLDPBD164456-250MGEA 22 EUR
BLDPBD164456-250MG BLDPBD164456-1G BLDPBD164456-5G BLDPBD164456-100MG
2-Amino-5-methoxypyrimidine 97%
2-Amino-5-methoxypyrimidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₇N₃O
Molecular Weight: 125,131 g/mol
Lagertemperatur: Ambient
MDL: MFCD00187864
CAS-Nummer: 13418-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR