Boc-(S)-3-amino-4-(3-trifluoromethylphenyl)butyric acid 98%

Synonyme: (S)-3-((tert-Butoxycarbonyl)amino)-4-(3-(trifluoromethyl)phenyl)butanoic acid

BD16440-1G BD16440-250MG BD16440-100MG
BLDPBD16440-1GEA 368 EUR
BLDPBD16440-1G BLDPBD16440-250MG BLDPBD16440-100MG
Boc-(S)-3-amino-4-(3-trifluoromethylphenyl)butyric acid 98%
Boc-(S)-3-amino-4-(3-trifluoromethylphenyl)butyric acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₆H₂₀F₃NO₄
Molecular Weight: 347,34 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD01861067
CAS-Nummer: 270065-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR