Boc-(R)-3-amino-4-(3-trifluoromethylphenyl)butyric acid 98%

Synonyme: (R)-3-((tert-Butoxycarbonyl)amino)-4-(3-(trifluoromethyl)phenyl)butanoic acid

BD16437-1G BD16437-100MG BD16437-250MG
BLDPBD16437-1GEA 471 EUR
BLDPBD16437-1G BLDPBD16437-100MG BLDPBD16437-250MG
Boc-(R)-3-amino-4-(3-trifluoromethylphenyl)butyric acid 98%
Boc-(R)-3-amino-4-(3-trifluoromethylphenyl)butyric acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₆H₂₀F₃NO₄
Molecular Weight: 347,34 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD01860969
CAS-Nummer: 269726-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR