2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile 95%

BD164228-1G BD164228-250MG BD164228-5G BD164228-25G
BLDPBD164228-1GEA 39 EUR
BLDPBD164228-1G BLDPBD164228-250MG BLDPBD164228-5G BLDPBD164228-25G
2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile 95%
2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₅ClF₃N
Molecular Weight: 219,5909 g/mol
Lagertemperatur: Ambient
MDL: MFCD04115850
CAS-Nummer: 886496-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR