2-Nitrobenzofuran 95%

BD164167-250MG BD164167-5G BD164167-100MG BD164167-1G
BLDPBD164167-250MGEA 29 EUR
BLDPBD164167-250MG BLDPBD164167-5G BLDPBD164167-100MG BLDPBD164167-1G
2-Nitrobenzofuran 95%
2-Nitrobenzofuran

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₅NO₃
Molecular Weight: 163,13 g/mol
Lagertemperatur: Ambient
MDL: MFCD00459786
CAS-Nummer: 33094-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR