4-(1-Aminoethyl)benzoic acid 95%

BLDPBD164159-100MGEA 57 EUR
BLDPBD164159-100MG BLDPBD164159-5G BLDPBD164159-250MG BLDPBD164159-1G
4-(1-Aminoethyl)benzoic acid 95%
4-(1-Aminoethyl)benzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832183
CAS-Nummer: 28357-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR