Ethyl morpholine-2-carboxylate 96%

BD164127-1G BD164127-250MG
BLDPBD164127-1GEA 93 EUR
BLDPBD164127-1G BLDPBD164127-250MG
Ethyl morpholine-2-carboxylate 96%
Ethyl morpholine-2-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₁₃NO₃
Lagertemperatur: Refrigerator
MDL: MFCD08751336
CAS-Nummer: 135782-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR