3,3',5,5'-Tetrabrom-2,2'-bithiophen 98%

BD164121-250MG BD164121-10G BD164121-5G BD164121-25G BD164121-1G BD164121-100G
BLDPBD164121-250MGEA 6 EUR
BLDPBD164121-250MG BLDPBD164121-10G BLDPBD164121-5G BLDPBD164121-25G BLDPBD164121-1G BLDPBD164121-100G
3,3',5,5'-Tetrabrom-2,2'-bithiophen 98%
3,3',5,5'-Tetrabrom-2,2'-bithiophen

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₈H₂Br₄S₂
Molecular Weight: 481,84 g/mol
Lagertemperatur: Freezer
MDL: MFCD00114807
CAS-Nummer: 125143-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR