Methyl 2-chloro-3-methylbenzoate 98%

BD164071-5G BD164071-25G BD164071-1G BD164071-100G
BLDPBD164071-5GEA 31 EUR
BLDPBD164071-5G BLDPBD164071-25G BLDPBD164071-1G BLDPBD164071-100G
Methyl 2-chloro-3-methylbenzoate 98%
Methyl 2-chloro-3-methylbenzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉ClO₂
Molecular Weight: 184,61 g/mol
Lagertemperatur: Ambient
MDL: MFCD11226202
CAS-Nummer: 920759-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR