2-Acetoxy-2',4'-difluoroacetophenone 97%

BD163953-250MG BD163953-100MG BD163953-1G
BLDPBD163953-250MGEA 15 EUR
BLDPBD163953-250MG BLDPBD163953-100MG BLDPBD163953-1G
2-Acetoxy-2',4'-difluoroacetophenone 97%
2-Acetoxy-2',4'-difluoroacetophenone

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₈F₂O₃
Molecular Weight: 214,17 g/mol
Lagertemperatur: Ambient
MDL: MFCD09743655
CAS-Nummer: 122263-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR