(2S,4R)-Methyl-4-(tert-butoxycarbonylamino)pyrrolidine-2-carboxylate 95%

Synonyme: (2S,4R)-Methyl 4-(tert-butoxycarbonylamino)pyrrolidine-2-carboxylate

BD163863-50MG BD163863-250MG BD163863-100MG
BLDPBD163863-50MGEA 93 EUR
BLDPBD163863-50MG BLDPBD163863-250MG BLDPBD163863-100MG
(2S,4R)-Methyl-4-(tert-butoxycarbonylamino)pyrrolidine-2-carboxylate 95%
(2S,4R)-Methyl-4-(tert-butoxycarbonylamino)pyrrolidine-2-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₁H₂₀N₂O₄
Molecular Weight: 244,29 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD08704530
CAS-Nummer: 473806-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR