1,5-Dibromnaphthalin 96%

BLDPBD163815-250MGEA 17 EUR
BLDPBD163815-250MG BLDPBD163815-25G BLDPBD163815-5G BLDPBD163815-10G BLDPBD163815-100G BLDPBD163815-1G
1,5-Dibromnaphthalin 96%
1,5-Dibromonaphthalene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₆Br₂
Molecular Weight: 285,9600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00089984
CAS-Nummer: 7351-74-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR