N-Boc-6-hydroxy-3-azabicyclo[3.2.0]heptane 98%

BLDPBD163806-100MGEA 137 EUR
BLDPBD163806-100MG
N-Boc-6-hydroxy-3-azabicyclo[3.2.0]heptane 98%
N-Boc-6-hydroxy-3-azabicyclo[3.2.0]heptane

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₉NO₃
Molecular Weight: 213,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12031304
CAS-Nummer: 663172-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR