6-Bromo-3-ethyl-2-methoxyquinoline 95%

BD163785-1G BD163785-100MG BD163785-250MG
BLDPBD163785-1GEA 230 EUR
BLDPBD163785-1G BLDPBD163785-100MG BLDPBD163785-250MG
6-Bromo-3-ethyl-2-methoxyquinoline 95%
6-Bromo-3-ethyl-2-methoxyquinoline

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₁₂BrNO
Molecular Weight: 266,13 g/mol
Lagertemperatur: Ambient
MDL: MFCD09029947
CAS-Nummer: 409346-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR