1-Boc-4-(aminomethyl)-4-hydroxypiperidine 95%

BLDPBD163778-1GEA 49 EUR
BLDPBD163778-1G BLDPBD163778-250MG BLDPBD163778-5G
1-Boc-4-(aminomethyl)-4-hydroxypiperidine 95%
1-Boc-4-(aminomethyl)-4-hydroxypiperidine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₁H₂₂N₂O₃
Molecular Weight: 230,3 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11579803
CAS-Nummer: 392331-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR