2-Amino-3-bromo-4-methylbenzoic acid 95%

BD163767-100MG BD163767-250MG BD163767-1G
BLDPBD163767-100MGEA 89 EUR
BLDPBD163767-100MG BLDPBD163767-250MG BLDPBD163767-1G
2-Amino-3-bromo-4-methylbenzoic acid 95%
2-Amino-3-bromo-4-methylbenzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₈BrNO₂
Molecular Weight: 230,05 g/mol
Lagertemperatur: Ambient
MDL: MFCD12755819
CAS-Nummer: 320740-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR