(S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid 95%

Synonyme: (S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid,(2S)-3-Amino-2-benzylpropanoic acid, N-BOC protected,(2S)-2-{[(tert-Butoxycarbonyl)amino]methyl}-3-phenylpropanoic acid

BD163752-100MG BD163752-1G BD163752-250MG
BLDPBD163752-100MGEA 138 EUR
BLDPBD163752-100MG BLDPBD163752-1G BLDPBD163752-250MG
(S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid 95%
(S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,332 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD07372495
CAS-Nummer: 189619-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR