trans-3-Aminocyclopentanol Hydrochlorid 95%

BD163722-250MG BD163722-1G BD163722-5G BD163722-100MG
BLDPBD163722-250MGEA 62 EUR
BLDPBD163722-250MG BLDPBD163722-1G BLDPBD163722-5G BLDPBD163722-100MG
trans-3-Aminocyclopentanol Hydrochlorid 95%
trans-3-Aminocyclopentanol Hydrochlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₁₂ClNO
Molecular Weight: 137,6079 g/mol
Lagertemperatur: Ambient
MDL: MFCD22380009
CAS-Nummer: 124555-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR