2-Amino-5-[(4-ethyl-1-piperazinyl)methyl]pyridine 97%

Synonyme: 5-((4-Ethylpiperazin-1-yl)methyl)pyridin-2-amine

BD163710-1G BD163710-250MG
BLDPBD163710-1GEA 43 EUR
BLDPBD163710-1G BLDPBD163710-250MG
2-Amino-5-[(4-ethyl-1-piperazinyl)methyl]pyridine 97%
2-Amino-5-[(4-ethyl-1-piperazinyl)methyl]pyridine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₂H₂₀N₄
Molecular Weight: 220,314 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD14706690
CAS-Nummer: 1180132-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR