4-(Benzyloxy)-3-ethoxybenzaldehyd 98%

Synonyme: 4-Benzyloxy-3-ethoxybenzaldehyde

BD16369-5G BD16369-1G BD16369-250MG BD16369-100MG
BLDPBD16369-5GEA 35 EUR
BLDPBD16369-5G BLDPBD16369-1G BLDPBD16369-250MG BLDPBD16369-100MG
4-(Benzyloxy)-3-ethoxybenzaldehyd 98%
4-(Benzyloxy)-3-ethoxybenzaldehyd

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₆H₁₆O₃
Molecular Weight: 256,3000 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00615145
CAS-Nummer: 60186-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR