3-Bromo-6-methylpyridazine 98%

BD163666-10G BD163666-250MG BD163666-5G BD163666-25G BD163666-1G
BLDPBD163666-10GEA 251 EUR
BLDPBD163666-10G BLDPBD163666-250MG BLDPBD163666-5G BLDPBD163666-25G BLDPBD163666-1G
3-Bromo-6-methylpyridazine 98%
3-Bromo-6-methylpyridazine

Storage condition: Inert atmosphere, 2-8°C

Formel: C5H5BrN2
Molecular Weight: 173,0106 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09831907
CAS-Nummer: 65202-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR