tert-Butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate 97%

BLDPBD163626-100MGEA 80 EUR
BLDPBD163626-100MG BLDPBD163626-1G BLDPBD163626-250MG
tert-Butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate 97%
tert-Butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₁H₂₂N₂O₃
Molecular Weight: 230,30 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13152268
CAS-Nummer: 1188265-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR