tert-Butyl 2,9-diazaspiro[5.5]undecane-2-carboxylate hydrochloride 97%

BD163622-1G BD163622-100MG BD163622-250MG
BLDPBD163622-1GEA 360 EUR
BLDPBD163622-1G BLDPBD163622-100MG BLDPBD163622-250MG
tert-Butyl 2,9-diazaspiro[5.5]undecane-2-carboxylate hydrochloride 97%
tert-Butyl 2,9-diazaspiro[5.5]undecane-2-carboxylate hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₂₇ClN₂O₂
Molecular Weight: 290,83 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD13185055
CAS-Nummer: 1023301-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR