N,N'-Bis(salicyliden)ethylendiamin 98%

BLDPBD16347-25GEA 27 EUR
BLDPBD16347-25G BLDPBD16347-10G BLDPBD16347-5G BLDPBD16347-500G BLDPBD16347-0.025G BLDPBD16347-100G
N,N'-Bis(salicyliden)ethylendiamin 98%
N,N'-Bis(salicyliden)ethylendiamin

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₁₆N₂O₂
Molecular Weight: 268,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002244
CAS-Nummer: 94-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR