6-Fluoro-1-methyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid 97%

BD16344-100MG
BLDPBD16344-100MGEA 167 EUR
BLDPBD16344-100MG
6-Fluoro-1-methyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid 97%
6-Fluoro-1-methyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₆H₁₆FN₃O₃S
Molecular Weight: 349,38 g/mol
Lagertemperatur: Ambient
MDL: MFCD00882994
CAS-Nummer: 112984-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR