Methyl 4,5-dichloro-3-hydroxythiophene-2-carboxylate 95%

BLDPBD163414-100MGEA 54 EUR
BLDPBD163414-100MG
Methyl 4,5-dichloro-3-hydroxythiophene-2-carboxylate 95%
Methyl 4,5-dichloro-3-hydroxythiophene-2-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₄Cl₂O₃S
Molecular Weight: 227,07 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD15071695
CAS-Nummer: 96232-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR