(3R,5S)-rel-3,5-Dimethylmorpholine 98%

BD163402-250MG BD163402-100MG BD163402-1G
BLDPBD163402-250MGEA 96 EUR
BLDPBD163402-250MG BLDPBD163402-100MG BLDPBD163402-1G
(3R,5S)-rel-3,5-Dimethylmorpholine 98%
(3R,5S)-rel-3,5-Dimethylmorpholine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₁₃NO
Molecular Weight: 115,17 g/mol
Lagertemperatur: Ambient
MDL: MFCD12405010
CAS-Nummer: 45597-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR