(3-Amino-5-chlorophenyl)methanol 95%

BD163346-250MG
BLDPBD163346-250MGEA 48 EUR
BLDPBD163346-250MG
(3-Amino-5-chlorophenyl)methanol 95%
(3-Amino-5-chlorophenyl)methanol
Formel: C₇H₈ClNO
Molecular Weight: 157,6 g/mol
MDL: MFCD11215511
CAS-Nummer: 79944-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR