4-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridine 98%

BD163299-100MG BD163299-250MG
BLDPBD163299-100MGEA 34 EUR
BLDPBD163299-100MG BLDPBD163299-250MG
4-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridine 98%
4-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridine
Formel: C₈H₈ClN
MDL: MFCD11113078
CAS-Nummer: 54664-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR