2-Hydroxy-3-methoxy-3,3-diphenylpropanoic acid 98%

BLDPBD163208-25GEA 183 EUR
BLDPBD163208-25G BLDPBD163208-5G BLDPBD163208-1G BLDPBD163208-10G
2-Hydroxy-3-methoxy-3,3-diphenylpropanoic acid 98%
2-Hydroxy-3-methoxy-3,3-diphenylpropanoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₁₆O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD13152279
CAS-Nummer: 178306-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR