1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride 95%

Synonyme: 1,4,5,6-Tetrahydro-pyrrolo[3,4-c]pyrazole,hydrochloride

BD163195-1G BD163195-250MG BD163195-5G
BLDPBD163195-1GEA 112 EUR
BLDPBD163195-1G BLDPBD163195-250MG BLDPBD163195-5G
1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride 95%
1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₉Cl₂N₃
Molecular Weight: 182,0511 g/mol
Lagertemperatur: Ambient
MDL: MFCD08457551
CAS-Nummer: 157327-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR