(3S,5S)-3,5-Dimethylmorpholine 98%

BD163192-250MG BD163192-100MG BD163192-1G
BLDPBD163192-250MGEA 108 EUR
BLDPBD163192-250MG BLDPBD163192-100MG BLDPBD163192-1G
(3S,5S)-3,5-Dimethylmorpholine 98%
(3S,5S)-3,5-Dimethylmorpholine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₁₃NO
Molecular Weight: 115,17 g/mol
Lagertemperatur: Ambient
MDL: MFCD14586370
CAS-Nummer: 154634-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR