Dipyridin-2-ylmethane 95%

BD163171-5G BD163171-250MG BD163171-1G BD163171-100MG
BLDPBD163171-5GEA 1470 EUR
BLDPBD163171-5G BLDPBD163171-250MG BLDPBD163171-1G BLDPBD163171-100MG
Dipyridin-2-ylmethane 95%
Dipyridin-2-ylmethane

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₀N₂
Molecular Weight: 170,21 g/mol
Lagertemperatur: Ambient
MDL: MFCD14706789
CAS-Nummer: 1132-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR