Dimethyl biphenyl-4,4'-dicarboxylate 98%

Synonyme: Dimethyl [1,1'-biphenyl]-4,4'-dicarboxylate,4,4'-Biphenyldicarboxylic Acid Dimethyl Ester,4,4'-Bibenzoic Acid Dimethyl Ester,Dimethyl-4,4'-biphenyldicarboxylate

BD16317-10G BD16317-500G BD16317-1G BD16317-100G BD16317-5G BD16317-25G
BLDPBD16317-10GEA 32 EUR
BLDPBD16317-10G BLDPBD16317-500G BLDPBD16317-1G BLDPBD16317-100G BLDPBD16317-5G BLDPBD16317-25G
Dimethyl biphenyl-4,4'-dicarboxylate 98%
Dimethyl biphenyl-4,4'-dicarboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₁₄O₄
Molecular Weight: 270,28 g/mol
Lagertemperatur: Ambient
MDL: MFCD00017201
CAS-Nummer: 792-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR