5-Fluoro-6-methylpyridine-2-carboxylic acid 97%

BD163156-5G BD163156-1G BD163156-250MG BD163156-100MG
BLDPBD163156-5GEA 667 EUR
BLDPBD163156-5G BLDPBD163156-1G BLDPBD163156-250MG BLDPBD163156-100MG
5-Fluoro-6-methylpyridine-2-carboxylic acid 97%
5-Fluoro-6-methylpyridine-2-carboxylic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₆FNO₂
Molecular Weight: 155,126 g/mol
Lagertemperatur: Ambient
MDL: MFCD11040267
CAS-Nummer: 1005474-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR