2,3-Dihydro-2-oxo-1H-benzimidazole-5-boronic acid, pinacol ester 95%

BLDPBD163071-1GEA 276 EUR
BLDPBD163071-1G BLDPBD163071-5G BLDPBD163071-100MG BLDPBD163071-250MG
2,3-Dihydro-2-oxo-1H-benzimidazole-5-boronic acid, pinacol ester 95%
2,3-Dihydro-2-oxo-1H-benzimidazole-5-boronic acid, pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₁₇BN₂O₃
Molecular Weight: 260,097 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09260491
CAS-Nummer: 710348-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR