2-Methyl-5-nitrophenylboronsäurepinakolester 98%

Synonyme: 2-Methyl-5-nitrophenylboronic acid, pinacol ester,2-Methyl-5-nitrophenylboronic Acid Pinacol Ester

BD162994-5G BD162994-25G BD162994-1G BD162994-10G BD162994-250MG
BLDPBD162994-5GEA 125 EUR
BLDPBD162994-5G BLDPBD162994-25G BLDPBD162994-1G BLDPBD162994-10G BLDPBD162994-250MG
2-Methyl-5-nitrophenylboronsäurepinakolester 98%
2-Methyl-5-nitrophenylboronsäurepinakolester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₁₈BNO₄
Molecular Weight: 263,1 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09027081
CAS-Nummer: 957062-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR