N-(3-Bromophenyl)-6,7-dimethoxyquinazolin-4-amine 99+%

Synonyme: Tyrphostin AG 1517

BD16294-250MG BD16294-100MG
BLDPBD16294-250MGEA 436 EUR
BLDPBD16294-250MG BLDPBD16294-100MG
N-(3-Bromophenyl)-6,7-dimethoxyquinazolin-4-amine 99+%
N-(3-Bromophenyl)-6,7-dimethoxyquinazolin-4-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₆H₁₄BrN₃O₂
Molecular Weight: 360,2 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00934896
CAS-Nummer: 153436-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR