1-(3-Aminophenyl)-3-methyl-2-imidazolidinon 98%

BLDPBD162909-1GEA 144 EUR
BLDPBD162909-1G BLDPBD162909-5G BLDPBD162909-100MG BLDPBD162909-250MG
1-(3-Aminophenyl)-3-methyl-2-imidazolidinon 98%
1-(3-Aminophenyl)-3-methyl-2-imidazolidinon

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₀H₁₃N₃O
Molecular Weight: 191,2300 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05864562
CAS-Nummer: 517918-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR