1-(4-Bromophenyl)cyclobutane-1-carbonitrile 95%

BLDPBD162906-250MGEA 41 EUR
BLDPBD162906-250MG BLDPBD162906-5G BLDPBD162906-1G BLDPBD162906-100MG
1-(4-Bromophenyl)cyclobutane-1-carbonitrile 95%
1-(4-Bromophenyl)cyclobutane-1-carbonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₁H₁₀BrN
Molecular Weight: 236,108 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10688823
CAS-Nummer: 485828-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR