Quinoline-4-carboxamide 98%

BLDPBD162899-1GEA 229 EUR
BLDPBD162899-1G BLDPBD162899-250MG BLDPBD162899-5G BLDPBD162899-100MG
Quinoline-4-carboxamide 98%
Quinoline-4-carboxamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₈N₂O
Molecular Weight: 172,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672908
CAS-Nummer: 4363-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR