2,4-Dichlor-7-methylchinazolin 97%

BD162864-1G BD162864-250MG BD162864-100MG
BLDPBD162864-1GEA 430 EUR
BLDPBD162864-1G BLDPBD162864-250MG BLDPBD162864-100MG
2,4-Dichlor-7-methylchinazolin 97%
2,4-Dichloro-7-methylquinazoline

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₆Cl₂N₂
Molecular Weight: 213,0633 g/mol
Lagertemperatur: Ambient
MDL: MFCD11858254
CAS-Nummer: 25171-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR