tert-Butyl (1-benzylazetidin-3-yl)carbamate 95%

BD162752-5G BD162752-1G BD162752-250MG BD162752-100MG
BLDPBD162752-5GEA 73 EUR
BLDPBD162752-5G BLDPBD162752-1G BLDPBD162752-250MG BLDPBD162752-100MG
tert-Butyl (1-benzylazetidin-3-yl)carbamate 95%
tert-Butyl (1-benzylazetidin-3-yl)carbamate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₂₂N₂O₂
Molecular Weight: 262,35 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09878667
CAS-Nummer: 1000577-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR