6-Nitro-3H-imidazo[4,5-b]pyridine 95%

BLDPBD162710-250MGEA 90 EUR
BLDPBD162710-250MG BLDPBD162710-1G BLDPBD162710-100MG
6-Nitro-3H-imidazo[4,5-b]pyridine 95%
6-Nitro-3H-imidazo[4,5-b]pyridine

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₄N₄O₂
Molecular Weight: 164,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11215559
CAS-Nummer: 3537-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR