tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate 98%

Synonyme: tert-Butyl 1-oxo-2,7-diazaspiro[4.5]decane-7-carboxylate

BD162611-1G BD162611-100MG BD162611-250MG
BLDPBD162611-1GEA 154 EUR
BLDPBD162611-1G BLDPBD162611-100MG BLDPBD162611-250MG
tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate 98%
tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₃H₂₂N₂O₃
Molecular Weight: 254,33 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11227060
CAS-Nummer: 923009-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR